Please login to your online account to display your discounted pricing

Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use, ACROS Organics™


Quantity 500mL

Manufacturer: thermo fisher scientific  AC383875000

 View more versions of this product

Catalog No. AC383875000

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Density 1g/cm3
Chemical Name or Material Buffer solution pH 9.2
MDL Number MFCD00163147
Name Note Borate buffer, traceable to NIST, ready to use
CAS 1330-43-4

Specifications

Chemical Name or Material Buffer solution pH 9.2
Name Note Borate buffer, traceable to NIST, ready to use
CAS 1330-43-4
MDL Number MFCD00163147