missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution pH 9.18 at 25°C, Certified, LabChem™
Supplier: LabChem LC124502
Specifications
Buffer Solution pH 9.18 at 25°C | |
Colorless | |
Boron | |
Liquid | |
Passes Test | |
Poly Bottle | |
Soluble in water | |
Certified |
1303-96-4,7732-18-5 | |
99.62,0.38 | |
9.18 ±0.01 | |
1 L | |
Na2B4O7·10H2O | |
Traceable to NIST | |
381.37 |