Please login to your online account to display your discounted pricing

Buffer Solution pH 9.0 at 25°C, Certified, LabChem™

$23.46 - $255.26

Chemical Identifiers

Chemical Name or Material Buffer Solution pH 9.0 at 25°C
CAS 7732-18-5
CAS Max % 99.27
CAS 7447-40-7
CAS Max % 0.37
CAS 10043-35-3
CAS Max % 0.31
CAS 1310-73-2
CAS Max % 0.05
Purity Grade Certified Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC124401

 
lab chem
LC124401
Poly Bottle 500mL Each for $23.46

LC124402

 
lab chem
LC124402
Poly Bottle 1L Each for $44.83

LC124404

 
lab chem
LC124404
Poly Bottle 4L Each for $110.20

LC124405

 
lab chem
LC124405
Cubitainer™ 20L Each for $255.26

LC124406

 
lab chem
LC124406
Cubitainer™ 10L Each for $180.00
Description & Specifications

Specifications

Formula Weight 74.55
Purity Grade Certified Grade
pH 9.00 ± 0.01
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor
Solubility Soluble in water
CAS 7447-40-7
Traceability to NIST Traceable to NIST
Quantity 500mL
CAS Max % 0.31
CAS Max % 0.37
Packaging Poly Bottle
CAS Max % 99.27
Recommended Storage Room Temperature
Color clear, colorless solution
CAS 7732-18-5
CAS Max % 0.05
Identification Passes Test
Chemical Name or Material Buffer Solution pH 9.0 at 25°C
Physical Form Liquid
Linear Formula KCl
Decomposition Information Boron; Hydrogen chloride
CAS 10043-35-3
CAS 1310-73-2
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Stor

GHS H Statement
May damage fertility or the unborn child.

Room Temperature