missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution pH 9.0 at 25°C, Certified, LabChem™
Supplier: LabChem LC124401
Specifications
Buffer Solution pH 9.0 at 25°C | |
99.27,0.37,0.31,0.05 | |
Boron; Hydrogen chloride | |
Liquid | |
Passes Test | |
Poly Bottle | |
Soluble in water | |
Certified |
Colorless | |
7447-40-7,10043-35-3,1310-73-2 | |
9.00 ±0.01 | |
500 mL | |
KCl | |
Traceable to NIST | |
74.55 |