missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution, pH 8.00 (Certified), Fisher Chemical™
Potassium Phosphate Monobasic/Sodium Hydroxide
$160.75 - $810.67
Specifications
Chemical Name or Material | Buffer Solution, pH 8.00 |
---|---|
CAS | 1310-73-2,7778-77-0,7732-18-5 |
pH | 8.0 |
Physical Form | Liquid |
Identification | Pass Test |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
SB112-500
|
Fisher Chemical
BPSB112500 |
500 mL | Poly Bottle |
Each for $160.75
Case of 6 Each for $964.50
|
|
||||
SB112-1
|
Fisher Chemical
FLSB1121 |
1 L | Poly Bottle |
Each for $183.42
Case of 6 Each for $1,100.52
|
|
||||
SB112-20
|
Fisher Chemical
BPSB11220 |
20 L | PolyPac™ |
Each for $810.67
|
|
||||
Specifications
Buffer Solution, pH 8.00 | |
8.0 | |
Pass Test | |
Traceable to NIST. |
1310-73-2,7778-77-0,7732-18-5 | |
Liquid | |
MFCD00147253 | |
Certified |