Please login to your online account to display your discounted pricing

Buffer Solution, pH 8.00 (Certified), Fisher Chemical


Potassium Phosphate Monobasic/Sodium Hydroxide

Quantity 1L
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLSB112-1

 View more versions of this product

Catalog No. SB112-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Buffer Solution, pH 8.00
CAS 1310-73-2
ChemAlert Storage Symbol Gray
Health Hazard 2 WARNING!
CAS 7778-77-0
Health Hazard 3 Emergency Overview
Irritating to eyes and skin. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:0
CAS 7732-18-5
Purity Grade Certified Grade
Physical Form Liquid
MDL Number MFCD00147253
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST Pass Test
NIST Traceable to NIST.
pH 8.0
Quantity 1L

Specifications

Chemical Name or Material Buffer Solution, pH 8.00
CAS 1310-73-2
CAS 7778-77-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00147253
Accessories
 

Emergency Overview
Irritating to eyes and skin. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents