missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution pH 8.0 at 25°C, Certified, LabChem™

Manufacturer:  LabChem LC124205

 View more versions of this product

Catalog No. LC124205



Specifications

Specifications

Buffer Solution pH 8.0 at 25°C
7778-77-0,1310-73-2
Passes Test
Liquid
Cubitainer™
Traceable to NIST
8.00 ±0.01
20L
99.2,0.68,0.12
KH2PO4
136.09
Certified
Soluble in water
Phosphorus oxides; Sodium oxide
Colorless
Documents
Promotions

Promotions