Please login to your online account to display your discounted pricing

Buffer solution, pH 8 (±0.01 at 25deg.C), No Color, NIST Traceable, Alfa Aesar™, Specpure™

Chemical Identifiers

Chemical Name or Material Buffer solution
CAS 1310-73-2
CAS 7778-77-0
CAS 7732-18-5
Physical Form Liquid
MDL Number MFCD00134130
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42420AP

  SDS
alfa aesar
42420AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42420K2

  SDS
alfa aesar
42420K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42420K7

  SDS
alfa aesar
42420K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Quantity 500mL
Color Yellow
Chemical Name or Material Buffer solution
MDL Number MFCD00134130
CAS 1310-73-2
Concentration or Composition (by Analyte or Components) Sodium hydroxide: 0.7%; Potassium dihydrogen phosphate: 0.7%; Water: 98.6%
Appearance Yellow liquid
CAS 7778-77-0
CAS 7732-18-5
Vapor Pressure 23 hPa (17mm Hg) at 20°C