missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 038711K2
Description
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
1336-21-6 | |
Liquid | |
H5NO | |
Yes | |
Miscible with water. | |
N.O | |
35.05 | |
CHEBI:18219 |
NIST Traceable | |
Green | |
1 L | |
MFCD00066650 | |
23 hPa (17mm Hg) at 20°C | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 | |
Specpure™ |
RUO – Research Use Only