missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer solution pH 7, Acros Organics

Chemical Identifiers

CAS 7732-18-5,7487-94-7,7778-77-0,7647-14-5
Chemical Name or Material Buffer solution pH 7
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC383970010 SDS Thermo Fisher Scientific
AC383970010
1 ea. N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

pH buffer
Description & Specifications

Chemical Identifiers

7732-18-5,7487-94-7,7778-77-0,7647-14-5
Buffer solution pH 7

Specifications

1 ea.
7487-94-7,7732-18-5,7647-14-5,7778-77-0
Buffer solution pH 7