Please login to your online account to display your discounted pricing

Buffer Solution pH 7.7 at 25°C, Certified, LabChem™

$40.23 - $318.06

Chemical Identifiers

Chemical Name or Material Buffer Solution pH 7.7 at 25°C
CAS 7732-18-5
CAS Max % 99.21
CAS 7778-77-0
CAS Max % 0.68
CAS 1310-73-2
CAS Max % 0.11
Purity Grade Certified Grade
Linear Formula KH2PO4
Formula Weight 136.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC124101

 
lab chem
LC124101
Poly Bottle 500mL Each for $40.23

LC124102

 
lab chem
LC124102
Poly Bottle 1L Each for $57.60

LC124104

 
lab chem
LC124104
Poly Bottle 4L Each for $154.06

LC124105

 
lab chem
LC124105
Cubitainer™ 20L Each for $318.06
Description & Specifications

Specifications

pH 7.70 ± 0.01
Purity Grade Certified Grade
Formula Weight 136.09
Chemical Name or Material Buffer Solution pH 7.7 at 25°C
Decomposition Information Phosphorus oxides; Sodium oxide
CAS Max % 0.11
Color Clear, colorless
Quantity 500mL
Physical Form Liquid
Linear Formula KH2PO4
CAS 7732-18-5
Identification Passes Test
CAS Max % 99.21
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 7778-77-0
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 0.68
Recommended Storage Room Temperature
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS 1310-73-2
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature