Please login to your online account to display your discounted pricing

Buffer Solution, pH 7.00, Color-Coded Yellow (Certified), Fisher Chemical


Quantity 20L
Packaging PolyPac™

Manufacturer: fisher scientific company  SB10720

 View more versions of this product

Catalog No. SB107-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Melting Point 0°C
Purity Grade Certified Grade
Quantity 20L
Physical Form Liquid
Solubility Information Soluble in water
NIST Traceable Traceable
pH Range 6.99 to 7.01 (at 25°C)
Density 1.0g/cm³
Specific Gravity 1
NIST Traceable to NIST.
pH 7.00
Color Yellow
Description Potassium Phosphate Monobasic/Sodium Hydroxide. pH/temperature conversions given on label.
Vapor Pressure 760mmHg at 20°C
CAS 7732-18-5
CAS 1934-21-0
Additional Information Vapor Pressure: 760mmHg at 20°C
CAS 1310-73-2
MDL Number MFCD00134130
ChemAlert Storage Symbol Gray
Health Hazard 2 CAUTION!
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Handle in accordance with good industrial hygiene and safety practice. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:0
Instability:0
Packaging PolyPac™
Absorbance Pass Test
Chemical Name or Material Buffer Solution, pH 7.00, Color-Coded Yellow
CAS 7778-77-0
Identification Pass Test
Traceability to NIST Pass Test

Specifications

Chemical Name or Material Buffer Solution, pH 7.00, Color-Coded Yellow
CAS 7778-77-0
CAS 1310-73-2
CAS 7732-18-5
CAS 1934-21-0
Purity Grade Certified Grade
MDL Number MFCD00134130
Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Handle in accordance with good industrial hygiene and safety practice. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!