Please login to your online account to display your discounted pricing

Buffer solution, pH 7.00 , Color-coded yellow, Certified, ACROS Organics™

$52.23 - $226.44

Chemical Identifiers

Chemical Name or Material Buffer solution pH 7.00
Name Note Color-coded yellow
CAS 7778-77-0
MDL Number MFCD00146206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC611060040

  SDS
fisher scientific acros
611060040
4L Each for $226.44

AC611065000

  SDS
fisher scientific acros
611065000
500mL Each for $52.23
Description & Specifications

Specifications

Quantity 4L
pH 6.99 to 7.01
Absorbance Pass Test
MDL Number MFCD00146206
Chemical Name or Material Buffer solution pH 7.00
Name Note Color-coded yellow
Additional Information Vapor Pressure: 760mmHg at 20°C
CAS 7778-77-0
Identification Pass Test