Please login to your online account to display your discounted pricing

Buffer Solution, pH 7.00, Color-Coded Green, Mettler Toledo™

$24.29 - $63.64

Chemical Identifiers

Chemical Name or Material Buffer Solution, pH 7.00
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

01-917-118

 
mettler toledo
ME63056125
500mL Each for $24.29

01-917-124

 
mettler toledo
ME63056124
4L Each for $63.64
Description & Specifications

Specifications

Quantity 500mL
Physical Form Liquid
NIST NIST Traceable
Chemical Name or Material Buffer Solution, pH 7.00