Please login to your online account to display your discounted pricing

Buffer Solution, pH 7.00 (Certified), Fisher Chemical


Potassium Phosphate Monobasic/Sodium Hydroxide

Quantity 10L
Packaging PolyPac™

Manufacturer: fisher scientific company  FLSB108-10

 View more versions of this product

Catalog No. SB108-10

  • / Each
 

Description & Specifications

Chemical Identifiers

Melting Point 0°C
Purity Grade Certified Grade
Quantity 10L
Density 1.003g/cm³
NIST Traceable to NIST.
pH 7.00
Color Colorless
Physical Form Liquid
Vapor Pressure 14mmHg at 20°C
MDL Number MFCD00134130
ChemAlert Storage Symbol Gray
CAS 1310-73-2
CAS 7732-18-5
Additional Information Vapor Pressure: 14mmHg at 20°C
CAS 7778-77-0
Synonym Chlorine Titrimeter Reagent
Identification Pass Test
Packaging PolyPac™
Chemical Name or Material Buffer Solution, pH 7.00

Specifications

Chemical Name or Material Buffer Solution, pH 7.00
CAS 7778-77-0
CAS 1310-73-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134130
Synonym Chlorine Titrimeter Reagent
Accessories
 

SDS & Safety Documents