Please login to your online account to display your discounted pricing

Buffer Solution pH 7.00 ±0.01 Yellow, EMD Millipore


Quantity 20L
Packaging

Manufacturer: emd millipore corporation  BX16323

 View more versions of this product

Catalog No. MBX16323

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 20L
Color Clear to yellow
Physical Form Liquid
Chemical Name or Material Buffer Solution pH 7.00 ±0.01
CAS 7558-79-4
Recommended Storage +15°C to +3°C
Linear Formula Mixture (Phosphate)
Packaging
Name Note Yellow

Specifications

Chemical Name or Material Buffer Solution pH 7.00 ±0.01
Name Note Yellow
CAS 7558-79-4
Physical Form Liquid
Linear Formula Mixture (Phosphate)
Accessories

+15°C to +3°C