missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution pH 7.00 ±0.01 Yellow, EMD Millipore

$39.57 - $207.49

Chemical Identifiers

CAS 7558-79-4
Physical Form Liquid
Linear Formula Mixture (Phosphate)
Chemical Name or Material Buffer Solution pH 7.00 ±0.01, Yellow
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MBX16323 Emd Millipore Corporation
BX16323
20L Each for $207.49
MBX16321 Emd Millipore Corporation
BX16321
Poly Bottle 500mL Each for $39.57
Specifications Description & Specifications

Chemical Identifiers

7558-79-4
Liquid
Mixture (Phosphate)
Buffer Solution pH 7.00 ±0.01, Yellow

Specifications

7558-79-4
Mixture (Phosphate)
Clear to yellow
Buffer Solution pH 7.00 ±0.01, Yellow
Liquid
20L

Safety and Handling

Recommended Storage : +15°C to +3°C