Please login to your online account to display your discounted pricing

Buffer solution, pH 7.00 (±0.01 at 25deg.C), No Color, NIST Traceable, Alfa Aesar™, Specpure™

$49.11 - $49.11

Chemical Identifiers

Chemical Name or Material Buffer solution
Purity Grade Specpure™
Physical Form Liquid
MDL Number MFCD00146206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38712K2

  SDS
alfa aesar
38712K2
1L Each for $49.11

AA38712AP

  SDS
alfa aesar
38712AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38712K7

  SDS
alfa aesar
38712K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Purity Grade Specpure™
Physical Form Liquid
Quantity 1L
Density 1.004-1.005g/cm3 at 20°C
Appearance Colorless liquid
Odor Odorless
Traceability to NIST Yes
Chemical Name or Material Buffer solution
MDL Number MFCD00146206