missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Buffer solution, pH 7.0, Coloured Yellow, Alfa Aesar™, Specpure™

Chemical Identifiers

CAS 1310-73-2,7778-77-0,7732-18-5
MDL Number MFCD00146252
Chemical Name or Material Buffer solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40439AP SDS Alfa Aesar™
40439AP
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA40439K2 SDS Alfa Aesar™
40439K2
1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA40439K7 SDS Alfa Aesar™
40439K7
4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1310-73-2,7778-77-0,7732-18-5
MFCD00146252
Buffer solution

Specifications

Liquid
500mL
Sodium hydroxide: 0.7%; Potassium dihydrogen phosphate: 0.7%; water: 98.6%
MFCD00146252
Buffer solution
Yellow
7778-77-0,1310-73-2,7732-18-5
Yellow liquid
23 hPa (17mm Hg) at 20°C