missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
$52.52 - $135.44
Specifications
Chemical Name or Material | Buffer solution |
---|---|
CAS | 1336-21-6 |
Color | Colorless |
Physical Form | Liquid |
Molecular Formula | H5NO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA42419AP
|
Thermo Scientific Chemicals
042419AP |
500 mL |
Each for $52.52
|
|
|||||
AA42419K2
|
Thermo Scientific Chemicals
042419K2 |
1 L |
Each for $83.84
|
|
|||||
AA42419K7
|
Thermo Scientific Chemicals
042419K7 |
4 L |
Each for $135.44
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
Colorless | |
H5NO | |
MFCD00066650 | |
N.O | |
14923 |
1336-21-6 | |
Liquid | |
Odorless | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
35.05 | |
CHEBI:18219 |
RUO – Research Use Only