missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution pH 6.0 at 25°C, Certified, LabChem™
Supplier: LabChem LC123302
Specifications
Buffer Solution pH 6.0 at 25°C | |
99.29,0.68,0.03 | |
Phosphorus oxides; Sodium oxide | |
Liquid | |
Passes Test | |
Poly Bottle | |
Soluble in water | |
Certified |
Colorless | |
7778-77-0,1310-73-2 | |
6.00 ±0.01 | |
1 L | |
KH2PO4 | |
Traceable to NIST | |
136.09 |