missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution pH 5.0 at 25°C, Certified, LabChem™
Supplier: LabChem LC123001
Specifications
Buffer Solution pH 5.0 at 25°C | |
Colorless | |
Formaldehyde; Carbon monoxide; Carbon dioxide | |
5.00 ±0.01 | |
500 mL | |
KHC8H4O4 | |
1g/mL | |
Soluble in water | |
Certified |
98.845,1.02,0.09,0.04,0.005 | |
877-24-7,1310-73-2,50-00-0,67-56-1 | |
1g/mL | |
Liquid | |
Passes Test | |
Poly Bottle | |
Traceable to NIST | |
204.23 |