Please login to your online account to display your discounted pricing

Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use, ACROS Organics™


Quantity 500mL

Manufacturer: thermo fisher scientific  AC383835000

 View more versions of this product

Catalog No. AC383835000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 204.22
Quantity 500mL
Chemical Name or Material Potassium hydrogen phthalate
Molecular Formula C8H5KO4
MDL Number MFCD00217672
CAS 877-24-7

Specifications

Chemical Name or Material Potassium hydrogen phthalate
CAS 877-24-7
Molecular Formula C8H5KO4
Formula Weight 204.22
MDL Number MFCD00217672