Please login to your online account to display your discounted pricing

Buffer solution pH 4, Phthalate buffer, traceable to NIST, colored red, ACROS Organics™

$53.43 - $53.43

Chemical Identifiers

Chemical Name or Material Buffer solution pH 4
Name Note Phthalate buffer, traceable to NIST
CAS 7732-18-5
CAS 7487-94-7
CAS 3244-88-0
MDL Number MFCD00217672
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383840025

  SDS
thermo fisher scientific
AC383840025
Plastic bottle 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC383840010

  SDS
thermo fisher scientific
AC383840010
Plastic bottle 1L Each for $53.43
Description & Specifications

Specifications

Physical Form Liquid
Quantity 2.5L
Color Clear red
Merck Index 14, 7612
Density 1.0000g/mL
NIST Traceable to NIST
pH 3.98 to 4.02 (20°C)
Name Note Phthalate buffer, traceable to NIST
MDL Number MFCD00217672
Packaging Plastic bottle
CAS 7487-94-7
Chemical Name or Material Buffer solution pH 4
CAS 3244-88-0
CAS 7732-18-5

pH buffer