Please login to your online account to display your discounted pricing

Buffer solution pH 4, Phthalate buffer, traceable to NIST, colored red, ACROS Organics™


Quantity 1L
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC383840010

 View more versions of this product

Catalog No. AC383840010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Quantity 1L
Color Clear red
pH 3.98 to 4.02 (20°C)
Density 1.0000g/mL
NIST Traceable to NIST
Merck Index 14, 7612
Chemical Name or Material Buffer solution pH 4
CAS 7732-18-5
MDL Number MFCD00217672
Packaging Plastic bottle
CAS 7487-94-7
CAS 3244-88-0
Name Note Phthalate buffer, traceable to NIST

Specifications

Chemical Name or Material Buffer solution pH 4
Name Note Phthalate buffer, traceable to NIST
CAS 7732-18-5
CAS 7487-94-7
CAS 3244-88-0
MDL Number MFCD00217672