Please login to your online account to display your discounted pricing

Buffer solution, pH 4.01 (±0.01 at 25deg.C), No Color, NIST Traceable, Alfa Aesar™, Specpure™

$48.07 - $48.07

Chemical Identifiers

Chemical Name or Material Buffer solution
Physical Form Liquid
MDL Number MFCD00146206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38714AP

  SDS
alfa aesar
38714AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38714K7

  SDS
alfa aesar
38714K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38714K2

  SDS
alfa aesar
38714K2
1L Each for $48.07
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Vapor Pressure 23 hPa (17mm Hg) at 20°C
MDL Number MFCD00146206
Chemical Name or Material Buffer solution
Appearance Colorless liquid
Odor Odorless