Please login to your online account to display your discounted pricing

Buffer solution, pH 4.00, color-coded red Certified, traceable to NIST, ACROS Organics™

$54.81 - $149.46

Chemical Identifiers

Chemical Name or Material Buffer solution, pH 4.00
Name Note Color-coded Red
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00146206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC611040040

  SDS
fisher scientific acros
611040040
4L Each for $149.46

AC611045000

  SDS
fisher scientific acros
611045000
500mL Each for $54.81
Description & Specifications

Specifications

Purity Grade Certified Grade
Quantity 4L
pH 3.99 to 4.01
Absorbance Pass Test
Chemical Name or Material Buffer solution, pH 4.00
Name Note Color-coded Red
MDL Number MFCD00146206
CAS 7732-18-5
Identification Pass Test