Please login to your online account to display your discounted pricing

Buffer Solution, pH 4.00, Color-Coded Red (Certified), Fisher Chemical


pH/temperature conversions given on label

Quantity 4L
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLSB1014

 View more versions of this product

Catalog No. SB101-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Melting Point 0°C
Purity Grade Certified Grade
Quantity 4L
Concentration or Composition (by Analyte or Components) Potassium Hydrogen Phthalate
Density 1.0g/cm³
Specific Gravity 1
Analyte or Component Names Potassium Hydrogen Phthalate
NIST Traceable to NIST.
pH 4.0
Color Red
Physical Form Liquid
Description pH/temperature conversions given on label.
Solubility Information Soluble in water
Absorbance Pass Test
CAS 7732-18-5
CAS 67-56-1
MDL Number MFCD00146206
ChemAlert Storage Symbol Gray
Health Hazard 2 CAUTION!
CAS 877-24-7
CAS 16423-68-0
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST Pass Test
CAS 50-00-0
Health Hazard 3 Emergency Overview
Contains a known or suspected carcinogen. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0
Chemical Name or Material Buffer Solution, pH 4.00, Color-Coded Red

Specifications

Chemical Name or Material Buffer Solution, pH 4.00, Color-Coded Red
CAS 50-00-0
CAS 67-56-1
CAS 877-24-7
CAS 7732-18-5
CAS 16423-68-0
Purity Grade Certified Grade
MDL Number MFCD00146206
Accessories
 

Emergency Overview
Contains a known or suspected carcinogen. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents