missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution, pH 4.00, Color-Coded Pink, Mettler Toledo™

$24.16 - $72.98

Chemical Identifiers

CAS 877-24-7
Physical Form Liquid
Molecular Formula HO2CC6H4CO2K
Formula Weight 204.23
Synonym Potassium Biphthalate
Chemical Name or Material Buffer Solution, pH 4.00
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-917-117 Mettler Toledo
ME63056121
500mL Each for $24.16
01-917-121 Mettler Toledo
ME63056120
4L Each for $72.98
Specifications Description & Specifications

Chemical Identifiers

877-24-7
HO2CC6H4CO2K
Potassium Biphthalate
Liquid
204.23
Buffer Solution, pH 4.00

Specifications

877-24-7
HO2CC6H4CO2K
Potassium Biphthalate
Buffer Solution, pH 4.00
Liquid
204.23
500mL