Please login to your online account to display your discounted pricing

Buffer Solution, pH 4.00 (Certified), Fisher Chemical


Quantity 20L
Packaging PolyPac™

Manufacturer: fisher scientific company  BPSB98-20

 View more versions of this product

Catalog No. SB98-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Buffer Solution, pH 4.00
CAS 877-24-7
ChemAlert Storage Symbol Gray
Health Hazard 2 CAUTION!
CAS 7732-18-5
Health Hazard 3 Emergency Overview
Contains a known or suspected carcinogen. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0
CAS 50-00-0
CAS 67-56-1
Purity Grade Certified Grade
Physical Form Liquid
MDL Number MFCD00146206
Identification Pass Test
Packaging PolyPac™
Traceability to NIST Pass Test
Density 1.0054g/cm³
Analyte or Component Names Potassium Acid Phthalate
NIST Traceable to NIST.
pH 4.0
Color Clear
Quantity 20L

Specifications

Chemical Name or Material Buffer Solution, pH 4.00
CAS 877-24-7
CAS 7732-18-5
CAS 50-00-0
CAS 67-56-1
Purity Grade Certified Grade
MDL Number MFCD00146206
Accessories
 

Emergency Overview
Contains a known or suspected carcinogen. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents