missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution, pH 4.00 (Certified), Fisher Chemical

Manufacturer:  Fisher Scientific Company BPSB98-10

 View more versions of this product

Catalog No. SB98-10


 

Specifications Description & Specifications

Specifications

Buffer Solution, pH 4.00
Certified Grade
MFCD00146206
PolyPac™
1.0054g/cm³
Traceable to NIST.
Clear
877-24-7,7732-18-5,50-00-0,67-56-1
Liquid
Pass Test
Pass Test
Potassium Acid Phthalate
4.0
10L

Chemical Identifiers

877-24-7,7732-18-5,50-00-0,67-56-1
MFCD00146206
Buffer Solution, pH 4.00
Certified Grade

Safety and Handling

SDS & Safety Documents

CAUTION!

Emergency Overview
Contains a known or suspected carcinogen. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0