missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution pH 4.0 at 25°C, Color Coded Red, Certified, LabChem™
Supplier: LabChem LC1228020
Specifications
Buffer Solution pH 4.0 at 25°C | |
98.935,1.02,0.04,0.005 | |
877-24-7,50-00-0,67-56-1 | |
1g/mL | |
Liquid | |
Passes Test | |
Pack | |
Traceable to NIST | |
204.23 |
Color Coded Red | |
Red | |
Formaldehyde; Carbon monoxide; Carbon dioxide | |
4.00 to 4.01 | |
20 x 20 mL | |
KHC8H4O4 | |
1g/mL | |
Soluble in water | |
Certified |