missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution, pH 3.5, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI147432

 View more versions of this product

Catalog No. 1474-32



Specifications

Specifications

Buffer Solution
7647-01-0,631-61-8,7732-18-5
Poly natural
1L
pH 3.5 Reagent
Liquid
Colorless
Documents