Please login to your online account to display your discounted pricing

Buffer Solution, pH 3.00 (Certified), Fisher Chemical


Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  BPSB97-500

 View more versions of this product

Catalog No. SB97-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Melting Point 0°C
Purity Grade Certified Grade
Quantity 500mL
Density 0.7g/cm³
Analyte or Component Names Potassium Acid Phthalate,Hydrochloric Acid
NIST Traceable to NIST.
pH 3.00
Color Clear
Physical Form Liquid
CAS 7647-01-0
Packaging Poly Bottle
CAS 50-00-0
MDL Number MFCD00146206
Health Hazard 2 CAUTION!
CAS 67-56-1
CAS 7732-18-5
Identification Pass Test
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0
Traceability to NIST Pass Test
Chemical Name or Material Buffer Solution, pH 3.00
CAS 877-24-7
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Buffer Solution, pH 3.00
CAS 877-24-7
CAS 7647-01-0
CAS 50-00-0
CAS 67-56-1
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00146206
Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents