Please login to your online account to display your discounted pricing

Buffer Solution, pH 3.00 (Certified), Fisher Chemical


Quantity 20L
Packaging PolyPac™

Manufacturer: fisher scientific company  BPSB97-20

 View more versions of this product

Catalog No. SB97-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Melting Point 0°C
Purity Grade Certified Grade
Quantity 20L
Density 0.7g/cm³
Analyte or Component Names Potassium Acid Phthalate,Hydrochloric Acid
NIST Traceable to NIST.
pH 3.00
Color Clear
Physical Form Liquid
Identification Pass Test
CAS 50-00-0
CAS 67-56-1
CAS 7732-18-5
ChemAlert Storage Symbol Gray
Health Hazard 2 CAUTION!
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0
Traceability to NIST Pass Test
MDL Number MFCD00146206
Packaging PolyPac™
Chemical Name or Material Buffer Solution, pH 3.00
CAS 877-24-7
CAS 7647-01-0

Specifications

Chemical Name or Material Buffer Solution, pH 3.00
CAS 877-24-7
CAS 7647-01-0
CAS 50-00-0
CAS 67-56-1
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00146206
Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents