missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution pH 3.0 at 25°C, Certified, LabChem™

Manufacturer:  LabChem LC122504

 View more versions of this product

Catalog No. LC122504



Specifications

Specifications

Buffer Solution pH 3.0 at 25°C
877-24-7,7647-01-0,50-00-0,67-56-1
Passes Test
Liquid
Poly Bottle
Traceable to NIST
1g/mL
3.00 ±0.01
4L
98.785,1.02,0.15,0.04,0.005
KHC8H4O4
204.23
Certified
Soluble in water
1g/mL
Formaldehyde; Carbon monoxide; Carbon dioxide
Colorless
Documents
Promotions

Promotions