Please login to your online account to display your discounted pricing

Buffer Solution, pH 2.00 (Certified), Fisher Chemical


Quantity 20L
Packaging PolyPac™

Manufacturer: fisher scientific company  BPSB96-20

 View more versions of this product

Catalog No. SB96-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Melting Point 0°C
Purity Grade Certified Grade
Quantity 20L
DOT Information DOT Class 8, : Corrosive
Density 1.0g/cm³
Analyte or Component Names Potassium Chloride,Hydrochloric Acid
NIST Traceable to NIST.
pH 2.00
Color Clear
Physical Form Liquid
CAS 7732-18-5
Identification In Pass Test
CAS 7647-01-0
CAS 50-00-0
CAS 67-56-1
MDL Number MFCD00146206
Health Hazard 2 CAUTION!
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. WARNING! This product contains a chemical known in the State of California to
Packaging PolyPac™
Traceability to NIST Pass Test
Chemical Name or Material Buffer Solution, pH 2.00
CAS 7447-40-7
ChemAlert Storage Symbol Gray
Health Hazard 1 Signal Word:CAUTION!

Specifications

Chemical Name or Material Buffer Solution, pH 2.00
CAS 7447-40-7
CAS 7647-01-0
CAS 50-00-0
CAS 67-56-1
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00146206
Accessories
 

Signal Word:CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. WARNING! This product contains a chemical known in the State of California to

CAUTION!