missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution, pH 2.00 (Certified), Fisher Chemical™
Supplier: Fisher Chemical BPSB961
Specifications
Buffer Solution, pH 2.00 | |
7447-40-7,7647-01-0,50-00-0,67-56-1,7732-18-5 | |
Undesignated | |
2.00 | |
1 L | |
MFCD00146206 | |
Traceable to NIST. |
0°C | |
100°C | |
1.0g/cm³ | |
Liquid | |
In Pass Test | |
Poly Bottle | |
Certified |