missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Buffer solution, pH 13.00 (+/-0.01 at 25°C), No Color, Specpure™, NIST Traceable

Manufacturer:  Alfa Aesar™ 46307K2

 View more versions of this product

Catalog No. AA46307K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Buffer solution, pH 13.00 (±0.01 at 25°C)
MFCD03458582
Specpure™
1L
NIST Traceable
Liquid
Colorless
Documents