Please login to your online account to display your discounted pricing

Buffer solution, pH 12.00 (+/-0.01 at 25°C), No Color, Specpure™, NIST Traceable, Alfa Aesar™

$32.26 - $32.26

Chemical Identifiers

Chemical Name or Material Buffer solution
Name Note No Color, Specpure™, NIST Traceable
MDL Number MFCD00147253
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA42425AP

  SDS
alfa aesar
42425AP
Each for $32.26
Description & Specifications

Specifications

pH 12.00 (± 0.01 at 25°C)
Quantity 500mL
Chemical Name or Material Buffer solution
TSCA TSCA
Name Note No Color, Specpure™, NIST Traceable
Physical Form Liquid
MDL Number MFCD00147253