Please login to your online account to display your discounted pricing

Buffer solution, pH 12 (±0.01 at 25deg.C), No Color, NIST Traceable, Alfa Aesar™, Specpure™

$49.57 - $90.19

Chemical Identifiers

Chemical Name or Material Buffer solution
Physical Form Liquid
MDL Number MFCD00147253
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42425K2

  SDS
alfa aesar
42425K2
1L Each for $49.57

AA42425K7

  SDS
alfa aesar
42425K7
4L Each for $90.19
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 1L
Density 1.004-1.005g/cm3 at 20°C
Chemical Name or Material Buffer solution
MDL Number MFCD00147253
Appearance Colorless liquid
Traceability to NIST Yes
Odor Odorless