missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer solution, pH 12.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 042425K7
Description
Buffer solution (pH 12.00) is used as a means of keeping the pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
Colorless | |
4 L | |
Odorless | |
Yes | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
14923 |
1336-21-6 | |
Liquid | |
H5NO | |
MFCD00066650 | |
Immiscible with water. | |
N.O | |
35.05 | |
CHEBI:18219 |
RUO – Research Use Only