missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution, pH 10, Color-coded blue, Certified, Thermo Scientific Chemicals
$111.86 - $195.27
Specifications
Absorbance | Pass Test |
---|---|
Chemical Name or Material | Buffer solution, pH 10.00 |
Name Note | Traceable to NIST |
pH | 9.98 to 10.02 |
Identification | Pass Test |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC611025000
|
Thermo Scientific Chemicals
611025000 |
500 mL |
Each for $111.86
|
|
|||||
AC611020040
|
Thermo Scientific Chemicals
611020040 |
4 L |
Each for $195.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pass Test | |
Traceable to NIST | |
Pass Test |
Buffer solution, pH 10.00 | |
9.98 to 10.02 | |
Certified |
RUO – Research Use Only