Please login to your online account to display your discounted pricing

Buffer solution pH 10, Borate buffer, traceable to NIST, ready to use, ACROS Organics™


Quantity 500mL

Manufacturer: thermo fisher scientific  AC383885000

 View more versions of this product

Catalog No. AC383885000

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Density 1g/cm3
MDL Number MFCD00163147
Chemical Name or Material Buffer solution pH 10
Name Note Borate buffer, traceable to NIST, ready to use

Specifications

Chemical Name or Material Buffer solution pH 10
Name Note Borate buffer, traceable to NIST, ready to use
MDL Number MFCD00163147