Please login to your online account to display your discounted pricing

Buffer solution pH 10, Borate buffer, traceable to NIST, ready to use, ACROS Organics™

$46.99 - $46.99

Chemical Identifiers

Chemical Name or Material Buffer solution pH 10
Name Note Borate buffer, traceable to NIST, ready to use
MDL Number MFCD00163147
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC383885000

  SDS
thermo fisher scientific
AC383885000
500mL Each for $46.99
Description & Specifications

Specifications

Quantity 500mL
Density 1g/cm3
Chemical Name or Material Buffer solution pH 10
MDL Number MFCD00163147
Name Note Borate buffer, traceable to NIST, ready to use