missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution, pH 10, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 258600010
Description
pH bufferSpecifications
Buffer solution pH 10 | |
Liquid | |
9.98 to 10.02 (20°C) | |
1L |
1332-57-6,144-55-8,7732-18-5 | |
Plastic bottle | |
Colorless |