missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution, pH 10, Thermo Scientific Chemicals
$81.39 - $258.81
Specifications
Chemical Name or Material | Buffer solution pH 10 |
---|---|
Color | Colorless |
CAS | 1332-57-6,144-55-8,7732-18-5 |
pH | 9.98 to 10.02 (20°C) |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC258605000
|
Thermo Scientific Chemicals
258605000 |
500 mL | Plastic bottle |
Each for $81.39
|
|
||||
AC258600025
|
Thermo Scientific Chemicals
258600025 |
2.5 L | Plastic bottle |
Each for $258.81
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH bufferSpecifications
Buffer solution pH 10 | |
1332-57-6,144-55-8,7732-18-5 | |
Liquid |
Colorless | |
9.98 to 10.02 (20°C) | |
Plastic bottle |
RUO – Research Use Only