Please login to your online account to display your discounted pricing

Buffer Solution, pH 10.00, Color-Coded Blue, Mettler Toledo™

$23.51 - $71.15

Chemical Identifiers

Chemical Name or Material Buffer Solution, pH 10.00
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

01-917-119

 
mettler toledo
ME63056129
500mL Each for $23.51

01-917-127

 
mettler toledo
ME63056128
4L Each for $71.15
Description & Specifications

Specifications

Chemical Name or Material Buffer Solution, pH 10.00
Physical Form Liquid
Quantity 500mL