missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer Solution, pH 10.00, Color-Coded Blue (Certified), Fisher Chemical™
Supplier: Fisher Chemical SB11520
Specifications
Pass Test | |
0°C | |
Blue | |
Potassium Carbonate/Potassium Tetraborate/ Potassium Hydroxide/Disodium EDTA Dihydrate. pH/temperature conversions given on label. | |
Liquid | |
Pass Test | |
PolyPac™ | |
Traceable to NIST. |
Buffer Solution, pH 10.00, Color-Coded Blue | |
100°C | |
6381-92-6,584-08-7,1332-77-0,1310-58-3 | |
10.0 | |
20 L | |
MFCD00146206 | |
1.013 at 25°C | |
Certified |