Please login to your online account to display your discounted pricing

Buffer Solution, pH 10.00 (Certified), Fisher Chemical


Potassium Carbonate/Potassium Tetraborate/Potassium Hydroxide/Disodium EDTA Dihydrate

Quantity 10L
Packaging PolyPac™

Manufacturer: fisher scientific company  FLSB116-10

 View more versions of this product

Catalog No. SB116-10

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Quantity 10L
Physical Form Liquid
pH 10
NIST Traceable to NIST.
Color Colorless
Traceability to NIST Pass Test
CAS 7732-18-5
CAS 584-08-7
Health Hazard 2 CAUTION!
MDL Number MFCD00146206
Packaging PolyPac™
Identification Pass Test
CAS 12228-88-5
CAS 1310-58-3
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Handle in accordance with good industrial hygiene and safety practice. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
ChemAlert Storage Symbol Gray
Chemical Name or Material Buffer Solution, pH 10.00
CAS 6381-92-6

Specifications

Chemical Name or Material Buffer Solution, pH 10.00
CAS 6381-92-6
CAS 584-08-7
CAS 1310-58-3
CAS 7732-18-5
CAS 12228-88-5
Purity Grade Certified Grade
MDL Number MFCD00146206
Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Handle in accordance with good industrial hygiene and safety practice. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents